2,4-Dimethylphenylhydrazinium chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 1-(2,4-Dimethylphenyl)hydrazine hydrochloride, 2,4-Dimethylphenylhydrazine hydrochloride

B21009.06 B21009.14 B21009.22
ALFAB21009.06EA 0 SGD
ALFAB21009.06 ALFAB21009.14 ALFAB21009.22
2,4-Dimethylphenylhydrazinium chloride ≥97%
2,4-Dimethylphenylhydrazinium chloride

Purity: 97% CAS No.: 60480-83-3 Molecular Formula: C8H12N2.HCl Formula Weight: 172.66 Melting Point: ca 188° dec. MDL No.: MFCD00013381

Formula: C₈H₁₃ClN₂
MW: 172,66 g/mol
Melting Pt: ∼ 188 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013381
CAS Number: 60480-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR