2,3,5,6-Tetrafluorophenylhydrazine ≥97%
Supplier: Thermo Scientific
|
Danger
|
B22600.06
B22600.14
ALFAB22600.06EA
0
SGD
ALFAB22600.06
ALFAB22600.14
2,3,5,6-Tetrafluorophenylhydrazine ≥97%
2,3,5,6-Tetrafluorophenylhydrazine
Purity: 97%
CAS No.: 653-11-2
Molecular Formula: C6H4F4N2
Formula Weight: 180.11
Storage Temperature: 2°C to 8°C
Melting Point: 86-92°
Beilstein Reference: 745777
MDL No.: MFCD00007575
Formula:
C₆H₄F₄N₂ Melting Pt: 86…92 °C |
MDL Number:
MFCD00007575 CAS Number: 653-11-2 EINECS: 211-494-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...