Methyl-3-methoxy-4-nitrobenzoate ≥98%

Supplier: Thermo Scientific

Synonyms: Methyl 3-methoxy-4-nitrobenzoate, 3-Methoxy-4-nitrobenzoic acid methyl ester, Methyl 3-methoxy-4-nitrobenzenecarboxylate

B23196.03 B23196.06
ALFAB23196.03EA 0 SGD
ALFAB23196.03 ALFAB23196.06
Methyl-3-methoxy-4-nitrobenzoate ≥98%
Methyl 3-methoxy-4-nitrobenzoate

Purity: 98% CAS No.: 5081-37-8 Molecular Formula: C9H9NO5 Formula Weight: 211.18 Melting Point: 83-87° Beilstein Reference: 2856207

Formula: C₉H₉NO₅
MW: 211,17 g/mol
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00051967
CAS Number: 5081-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR