2-Hydroxy-4-(trifluoromethyl)pyridine ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(Trifluoromethyl)pyridin-2-ol
H25756.MD
H25756.06
ALFAH25756.MDEA
0
SGD
ALFAH25756.MD
ALFAH25756.06
2-Hydroxy-4-(trifluoromethyl)pyridine ≥98%
2-Hydroxy-4-(trifluoromethyl)pyridine
Purity: 98%
CAS No.: 50650-59-4
Molecular Formula: C6H4F3NO
Formula Weight: 163.10
Melting Point: 158-160°
MDL No.: MFCD00153195
Formula:
C₆H₄F₃NO MW: 163,1 g/mol Melting Pt: 158…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153195 CAS Number: 50650-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...