Methyl-2-fluoro-6-methoxybenzoate ≥98%

Supplier: Thermo Scientific

Synonyms: Methyl 2-fluoro-6-methoxybenzoate, 2-Fluoro-6-methoxybenzoic acid methyl ester

H32955.03 H32955.09
ALFAH32955.03EA 0 SGD
ALFAH32955.03 ALFAH32955.09
Methyl-2-fluoro-6-methoxybenzoate ≥98%
Methyl 2-fluoro-6-methoxybenzoate

Purity: 98% CAS No.: 178747-79-0 Molecular Formula: C9H9FO3 Formula Weight: 184.17 Refractive Index: 1.5010

Formula: C₉H₉FO₃
MW: 184,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788537
CAS Number: 178747-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR