2,5-Dimethylphenylhydrazinium chloride ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 1-(2,5-Dimethylphenyl)hydrazine hydrochloride, 2,5-Dimethylphenylhydrazine hydrochloride

L12496.06 L12496.14
ALFAL12496.06EA 0 SGD
ALFAL12496.06 ALFAL12496.14
2,5-Dimethylphenylhydrazinium chloride ≥98%
2,5-Dimethylphenylhydrazinium chloride

Purity: 98% CAS No.: 56737-78-1 Molecular Formula: C8H12N2.HCl Formula Weight: 172.66 Melting Point: ca 205° dec. Beilstein Reference: 6118180

Formula: C₈H₁₃ClN₂
MW: 172,66 g/mol
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013382
CAS Number: 56737-78-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR