3-Bromobenzaldehyde diethyl acetal ≥97%
Supplier: Thermo Scientific
Synonyms:
3-Bromo-α,α-diethoxytoluene, 3-Bromobenzaldehyde Diethyl Acetal, 1-Bromo-3-(diethoxymethyl)benzene
L15514.06
L15514.14
L15514.22
ALFAL15514.06EA
0
SGD
ALFAL15514.06
ALFAL15514.14
ALFAL15514.22
3-Bromobenzaldehyde diethyl acetal ≥97%
3-Bromobenzaldehyde diethyl acetal
Purity: 97%
CAS No.: 75148-49-1
Molecular Formula: C11H15BrO2
Formula Weight: 259.14
Flash Point: 104°(219°F)
Refractive Index: 1.5140
Boiling Point: 88-91°/0.8mm
Beilstein Reference: 6392356
MDL No.: MFCD01075697
Formula:
C₁₁H₁₅BrO₂ MW: 259,14 g/mol Boiling Pt: 88…91 °C (0,8 mmHg) Density: 1,271 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01075697 CAS Number: 75148-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...