2-Hydroxy-5-methylpyridine ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Methyl-2-pyridone, 2-hydroxy-5-picoline

L19528.06 L19528.14
ALFAL19528.06EA 170 SGD
ALFAL19528.06 ALFAL19528.14
2-Hydroxy-5-methylpyridine ≥98%
2-Hydroxy-5-methylpyridine

Purity: 98% CAS No.: 1003-68-5 Molecular Formula: C6H7NO Formula Weight: 109.13 Odour: Odourless Melting Point: 183-187° Beilstein Reference: 107074 MDL No.: MFCD03092888

Formula: C₆H₇NO
MW: 109,13 g/mol
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD03092888
CAS Number: 1003-68-5
EINECS: 213-719-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR