2-Bromo-4-fluorobenzonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
L20115.03
L20115.06
L20115.14
ALFAL20115.03EA
0
SGD
ALFAL20115.03
ALFAL20115.06
ALFAL20115.14
2-Bromo-4-fluorobenzonitrile ≥98%
2-Bromo-4-fluorobenzonitrile
Purity: 98%
CAS No.: 36282-26-5
Molecular Formula: C7H3BrFN
Formula Weight: 200.01
Melting Point: 74-78°
Beilstein Reference: 1940597
UN No.: 3439
MDL No.: MFCD00672924
Formula:
C₇H₃BrFN MW: 200,01 g/mol Melting Pt: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672924 CAS Number: 36282-26-5 EINECS: 252-949-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...