1-Bromo-3-(trifluoromethoxy)benzene ≥98%
Supplier: Thermo Scientific
|
Warning
|
A16561.06
A16561.14
ALFAA16561.06EA
0
SGD
ALFAA16561.06
ALFAA16561.14
1-Bromo-3-(trifluoromethoxy)benzene ≥98%
1-Bromo-3-(trifluoromethoxy)benzene
Purity: 98+%
CAS No.: 2252-44-0
Molecular Formula: C7H4BrF3O
Formula Weight: 241.01
Flash Point: 62°(143°F)
Refractive Index: 1.4620
Boiling Point: 156-158°
Beilstein Reference: 1945938
MDL No.: MFCD00040943
Formula:
C₇H₄BrF₃O Boiling Pt: 156…158 °C Density: 1,620 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040943 CAS Number: 2252-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...