2,4-Dibromoanisole ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,3-Dibromo-4-methoxybenzene, 2,4-Dibromo-1-methoxybenzene
A16200.14
A16200.22
ALFAA16200.14EA
0
SGD
ALFAA16200.14
ALFAA16200.22
2,4-Dibromoanisole ≥98%
2,4-Dibromoanisole
Purity: 98+%
CAS No.: 21702-84-1
Molecular Formula: C7H6Br2O
Formula Weight: 265.93
Melting Point: 60-64°
Boiling Point: 82-88°/0.3mm
MDL No.: MFCD00000079
Formula:
C₇H₆Br₂O MW: 265,93 g/mol Boiling Pt: 82…88 °C (0,3 mmHg) Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000079 CAS Number: 21702-84-1 EINECS: 244-536-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...