3-Bromothiophenol ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
m-Bromothiophenol, 3-Bromobenzenethiol, 1-Bromo-3-mercaptobenzene, 3-Bromobenzene-1-thiol
B25092.06
B25092.03
B25092.14
ALFAB25092.06EA
0
SGD
ALFAB25092.06
ALFAB25092.03
ALFAB25092.14
3-Bromothiophenol ≥97%
3-Bromothiophenol
Purity: 97%
CAS No.: 6320-01-0
Molecular Formula: C6H5BrS
Formula Weight: 189.08
Flash Point: 105°(221°F)
Refractive Index: 1.6324
Odour: Unpleasant
Boiling Point: 119-121°/21mm
Beilstein Reference: 1926242
MDL No.: MFCD00009603
Formula:
C₆H₅BrS MW: 189,08 g/mol Boiling Pt: 119…121 °C (21 mmHg) Density: 1,599 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009603 CAS Number: 6320-01-0 EINECS: 228-664-9 UN: 3334 ADR: 9, |
Learn more
![](/m-sg.vwr.com/en_SG/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...