4-Methoxysalicylaldehyde ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Hydroxy-4-methoxybenzaldehyde

A12971.03 A12971.06 A12971.14
ALFAA12971.03EA 0 SGD
ALFAA12971.03 ALFAA12971.06 ALFAA12971.14
4-Methoxysalicylaldehyde ≥98%
4-Methoxysalicylaldehyde

Purity: 98% CAS No.: 673-22-3 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 110°(230°F) Melting Point: 40-44° Beilstein Reference: 1072443 MDL No.: MFCD00003327

Formula: C₈H₈O₃
MW: 152,15 g/mol
Boiling Pt: 124 °C (12 mmHg)
Melting Pt: 40…44 °C
Density: 1,2065 g/cm³ (51 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003327
CAS Number: 673-22-3
EINECS: 211-604-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR