2-(Dimethylamino)benzoic acid 98%
Supplier: Thermo Scientific
|
Warning
|
A12967.03
A12967.06
A12967.14
ALFAA12967.03EA
0
SGD
ALFAA12967.03
ALFAA12967.06
ALFAA12967.14
2-(Dimethylamino)benzoic acid 98%
2-(Dimethylamino)benzoic acid
Purity: 98%
CAS No.: 610-16-2
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Odour: Odourless
Melting Point: 70-74°
Beilstein Reference: 2089466
MDL No.: MFCD00016496
Formula:
C₉H₁₁NO₂ MW: 165,19 g/mol Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016496 CAS Number: 610-16-2 EINECS: 210-209-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...