2-Chloroveratraldehyde ≥98%

Supplier: Thermo Scientific

Synonyms: 2-Chloro-3,4-dimethoxybenzaldehyde

A10623.06 A10623.14
ALFAA10623.06EA 0 SGD
ALFAA10623.06 ALFAA10623.14
2-Chloroveratraldehyde ≥98%
2-Chloroveratraldehyde

Purity: 98% CAS No.: 5417-17-4 Molecular Formula: C9H9ClO3 Formula Weight: 200.62 Melting Point: 70-72° Beilstein Reference: 2109344

Formula: C₉H₉ClO₃
MW: 200,62 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00084942
CAS Number: 5417-17-4
EINECS: 226-515-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR