3,5-Dichlorobenzonitrile ≥98%

Supplier: Thermo Scientific
Danger

A11213.14 A11213.03 A11213.06
ALFAA11213.14EA 0 SGD
ALFAA11213.14 ALFAA11213.03 ALFAA11213.06
3,5-Dichlorobenzonitrile ≥98%
3,5-Dichlorobenzonitrile

Purity: 98% CAS No.: 6575-00-4 Molecular Formula: C7H3Cl2N Formula Weight: 172.01 Melting Point: 64-68° Beilstein Reference: 2207019 UN No.: 3439 MDL No.: MFCD00001800

Formula: C₇H₃Cl₂N
MW: 172,01 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00001800
CAS Number: 6575-00-4
EINECS: 229-495-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR