3,5-Dichloroanthranilic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-3,5-dichlorobenzoic acid

A16969.09 A16969.18 A16969.30
ALFAA16969.09EA 0 SGD
ALFAA16969.09 ALFAA16969.18 ALFAA16969.30
3,5-Dichloroanthranilic acid 97%
3,5-Dichloroanthranilic acid

Purity: 97% CAS No.: 2789-92-6 Molecular Formula: C7H5Cl2NO2 Formula Weight: 206.03 Melting Point: 225-227° Beilstein Reference: 779100

Formula: C₇H₅Cl₂NO₂
MW: 206,03 g/mol
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 225…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00017088
CAS Number: 2789-92-6
EINECS: 220-520-3
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR