4-Amino-2-chlorobenzoic acid 98%

Supplier: Thermo Scientific
Warning

A10116.06 A10116.14 A10116.22
ALFAA10116.06EA 0 SGD
ALFAA10116.06 ALFAA10116.14 ALFAA10116.22
4-Amino-2-chlorobenzoic acid 98%
4-Amino-2-chlorobenzoic acid

Purity: 98% CAS No.: 2457-76-3 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 220° dec. Beilstein Reference: 2803668

Formula: C₇H₆ClNO₂
MW: 171,58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: ∼ 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007772
CAS Number: 2457-76-3
EINECS: 219-540-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR