4-Chloro-3,5-dinitrobenzoic acid ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3,5-Dinitro-4-chlorobenzoic acid
A12373.14
A12373.22
A12373.36
ALFAA12373.14EA
0
SGD
ALFAA12373.14
ALFAA12373.22
ALFAA12373.36
4-Chloro-3,5-dinitrobenzoic acid ≥99%
4-Chloro-3,5-dinitrobenzoic acid
Purity: 99%
CAS No.: 118-97-8
Molecular Formula: C7H3ClN2O6
Formula Weight: 246.56
Flash Point: 186°(367°F)
Melting Point: 159-162°
Boiling Point: 315°
Beilstein Reference: 668253
MDL No.: MFCD00007080
Formula:
C₇H₃ClN₂O₆ MW: 246,56 g/mol Boiling Pt: 315 °C Melting Pt: 159…162 °C Flash Pt: 186 °C (367 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007080 CAS Number: 118-97-8 EINECS: 204-290-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...