1,2-Dichloro-4-fluorobenzene ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3,4-Dichlorofluorobenzene
A11357.18
A11357.09
ALFAA11357.18EA
0
SGD
ALFAA11357.18
ALFAA11357.09
1,2-Dichloro-4-fluorobenzene ≥98%
1,2-Dichloro-4-fluorobenzene
Purity: 98+%
CAS No.: 1435-49-0
Molecular Formula: C6H3Cl2F
Formula Weight: 165.00
Flash Point: 64°(147°F)
Refractive Index: 1.5240
Melting Point: -1°
Boiling Point: 168-171°
Beilstein Reference: 1861278
MDL No.: MFCD00018119
Formula:
C₆H₃Cl₂F MW: 164,99 g/mol Boiling Pt: 168…171 °C Melting Pt: –1 °C Density: 1,403 Flash Pt: 64 °C (147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00018119 CAS Number: 1435-49-0 EINECS: 215-858-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...