1,4-Dichloro-2-fluorobenzene ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 2,5-Dichlorofluorobenzene

B21282.06 B21282.14
ALFAB21282.06EA 0 SGD
ALFAB21282.06 ALFAB21282.14
1,4-Dichloro-2-fluorobenzene ≥99%
1,4-Dichloro-2-fluorobenzene

Purity: 99% CAS No.: 348-59-4 Molecular Formula: C6H3Cl2F Formula Weight: 165.00 Flash Point: 65°(149°F) Refractive Index: 1.5245 Melting Point: 4° Boiling Point: 168° Beilstein Reference: 2355590 MDL No.: MFCD00060656

Formula: C₆H₃Cl₂F
MW: 164,99 g/mol
Boiling Pt: 168 °C
Melting Pt: 4 °C
Density: 1,383
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060656
CAS Number: 348-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR