4-Chloro-2-fluorobenzoic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
B25277.03
B25277.06
B25277.14
ALFAB25277.03EA
0
SGD
ALFAB25277.03
ALFAB25277.06
ALFAB25277.14
4-Chloro-2-fluorobenzoic acid ≥98%
4-Chloro-2-fluorobenzoic acid
Purity: 98%
CAS No.: 446-30-0
Molecular Formula: C7H4ClFO2
Formula Weight: 174.56
Melting Point: 203-206°
MDL No.: MFCD00042468
Formula:
C₇H₄ClFO₂ MW: 174,56 g/mol Melting Pt: 203…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042468 CAS Number: 446-30-0 EINECS: 207-162-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...