2,4,5-Trifluoroaniline ≥98%

Supplier: Thermo Scientific
Danger

A16765.06 A16765.14
ALFAA16765.06EA 0 SGD
ALFAA16765.06 ALFAA16765.14
2,4,5-Trifluoroaniline ≥98%
2,4,5-Trifluoroaniline

Purity: 98% CAS No.: 367-34-0 Molecular Formula: C6H4F3N Formula Weight: 147.10 Melting Point: 59-63° Beilstein Reference: 2085734 UN No.: 2811 MDL No.: MFCD00007649

Formula: C₆H₄F₃N
MW: 147,1 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00007649
CAS Number: 367-34-0
EINECS: 206-692-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR