1,2,3-Trifluoro-4-nitrobenzene ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,3,4-Trifluoronitrobenzene
A16313.06
A16313.14
ALFAA16313.06EA
0
SGD
ALFAA16313.06
ALFAA16313.14
1,2,3-Trifluoro-4-nitrobenzene ≥97%
1,2,3-Trifluoro-4-nitrobenzene
Purity: 97%
CAS No.: 771-69-7
Molecular Formula: C6H2F3NO2
Formula Weight: 177.08
Flash Point: 93°(199°F)
Refractive Index: 1.4920
Boiling Point: 91-93°/20mm
Beilstein Reference: 2449746
UN No.: 2810
MDL No.: MFCD00041546
Formula:
C₆H₂F₃NO₂ MW: 177,08 g/mol Boiling Pt: 91…93 °C (20 mmHg) Density: 1,541 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041546 CAS Number: 771-69-7 EINECS: 212-238-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...