1,2,4-Trifluoro-5-nitrobenzene ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 2,4,5-Trifluoronitrobenzene

A16579.03 A16579.06 A16579.14
ALFAA16579.03EA 0 SGD
ALFAA16579.03 ALFAA16579.06 ALFAA16579.14
1,2,4-Trifluoro-5-nitrobenzene ≥99%
1,2,4-Trifluoro-5-nitrobenzene
Formula: C₆H₂F₃NO₂
MW: 177,08 g/mol
Boiling Pt: 195 °C
Melting Pt: –11 °C
Density: 1,544
Flash Pt: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007051
CAS Number: 2105-61-5
EINECS: 218-281-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR