1,3,5-Trifluoro-2-nitrobenzene ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Nitro-2,4,6-trifluorobenzene, 2,4,6-Trifluoronitrobenzene

A17035.03 A17035.06 A17035.14
ALFAA17035.03EA 0 SGD
ALFAA17035.03 ALFAA17035.06 ALFAA17035.14
1,3,5-Trifluoro-2-nitrobenzene ≥98%
1,3,5-Trifluoro-2-nitrobenzene

Purity: 98% CAS No.: 315-14-0 Molecular Formula: C6H2F3NO2 Formula Weight: 177.08 Flash Point: 77°(170°F) Refractive Index: 1.4790 Melting Point: 2-5° Boiling Point: 171-172° Beilstein Reference: 2106869 UN No.: 2810 MDL No.: MFCD00014687

Formula: C₆H₂F₃NO₂
MW: 177,08 g/mol
Boiling Pt: 171…172 °C
Melting Pt: 2…5 °C
Density: 1,514
Flash Pt: 77 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014687
CAS Number: 315-14-0
EINECS: 206-248-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR