3-Iodobenzonitrile ≥99%

Supplier: Thermo Scientific
Danger

B22020.09 B22020.18 B22020.88
ALFAB22020.09EA 0 SGD
ALFAB22020.09 ALFAB22020.18 ALFAB22020.88
3-Iodobenzonitrile ≥99%
3-Iodobenzonitrile

Purity: 99% CAS No.: 69113-59-3 Molecular Formula: C7H4IN Formula Weight: 229.02 Flash Point: 110°(230°F) Melting Point: 38-42° UN No.: 3439 MDL No.: MFCD00079762

Formula: C₇H₄IN
MW: 229,02 g/mol
Melting Pt: 38…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00079762
CAS Number: 69113-59-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR