1,2-Diethoxybenzene ≥98%

Supplier: Thermo Scientific
Warning

A17283.30 A17283.18
ALFAA17283.30EA 0 SGD
ALFAA17283.30 ALFAA17283.18
1,2-Diethoxybenzene ≥98%
1,2-Diethoxybenzene

Purity: 98% CAS No.: 2050-46-6 Molecular Formula: C10H14O2 Formula Weight: 166.22 Melting Point: 43-45° Boiling Point: 218-220° MDL No.: MFCD00015143

Formula: C₁₀H₁₄O₂
MW: 166,22 g/mol
Boiling Pt: 218…220 °C
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00015143
CAS Number: 2050-46-6
EINECS: 218-089-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR