3-(3-Chlorophenyl)propionic acid 96%

Supplier: Thermo Scientific
Danger

H33266.03 H33266.06 H33266.14
ALFAH33266.03EA 0 SGD
ALFAH33266.03 ALFAH33266.06 ALFAH33266.14
3-(3-Chlorophenyl)propionic acid 96%
3-(3-Chlorophenyl)propionic acid

Purity: 96% CAS No.: 21640-48-2 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 MDL No.: MFCD01310801

Formula: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01310801
CAS Number: 21640-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR