Methyl-4-chlorobutyrate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 4-chlorobutyrate

L03734.14 L03734.22
ALFAL03734.14EA 66.3 SGD
ALFAL03734.14 ALFAL03734.22
Methyl-4-chlorobutyrate ≥98%
Methyl 4-chlorobutyrate

Purity: 98% CAS No.: 3153-37-5 Molecular Formula: Cl(CH2)3CO2CH3 Formula Weight: 136.58 Flash Point: 59°(138°F) Refractive Index: 1.4320 Boiling Point: 175-176° Beilstein Reference: 1745117 UN No.: 3272 MDL No.: MFCD00001003

Formula: C₅H₉ClO₂
Boiling Pt: 175…176 °C
Density: 1,125
Flash Pt: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001003
CAS Number: 3153-37-5
EINECS: 221-592-9
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR