6-Fluorosalicylaldehyde ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-6-hydroxybenzaldehyde
L19900.03
L19900.MD
ALFAL19900.03EA
533
SGD
ALFAL19900.03
ALFAL19900.MD
6-Fluorosalicylaldehyde ≥97%
6-Fluorosalicylaldehyde
Purity: 97%
CAS No.: 38226-10-7
Molecular Formula: C7H5FO2
Formula Weight: 140.12
Melting Point: 37-39°
Beilstein Reference: 1935289
Formula:
C₇H₅FO₂ MW: 140,11 g/mol Melting Pt: 37…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD01090996 CAS Number: 38226-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...