3-Methyl-5-(trifluoromethyl)benzonitrile ≥97%

Supplier: Thermo Scientific
Danger

H32271.03 H32271.09
ALFAH32271.03EA 0 SGD
ALFAH32271.03 ALFAH32271.09
3-Methyl-5-(trifluoromethyl)benzonitrile ≥97%
3-Methyl-5-(trifluoromethyl)benzonitrile

Purity: 97% CAS No.: 261952-04-9 Molecular Formula: C9H6F3N Formula Weight: 185.15 Refractive Index: 1.4630

Formula: C₉H₆F₃N
Storage Temperature: Ambient
MDL Number: MFCD01631595
CAS Number: 261952-04-9
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR