5-Benzyloxytryptamine hydrochloride 98%

Supplier: Thermo Scientific

Synonyms: 2-[5-(Benzyloxy)-1H-indol-3-yl]ethanamine hydrochloride, 5-(phenylmethoxy)indole-3-ethanamine monohydrochloride

H51089.MD H51089.03
ALFAH51089.MDEA 140 SGD
ALFAH51089.MD ALFAH51089.03
5-Benzyloxytryptamine hydrochloride 98%
5-Benzyloxytryptamine hydrochloride

Purity: 98% CAS No.: 52055-23-9 Molecular Formula: C17H18N2O.HCl Formula Weight: 302.80 Melting Point: 259-263° MDL No.: MFCD00012685

Formula: C₁₇H₁₉ClN₂O
Melting Pt: 259…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00037972
CAS Number: 52055-23-9
EINECS: 257-626-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR