4-Nitrobenzaldehyde oxime ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Nitrobenzaldoxime

A15525.06 A15525.14
ALFAA15525.06EA 0 SGD
ALFAA15525.06 ALFAA15525.14
4-Nitrobenzaldehyde oxime ≥98%
4-Nitrobenzaldehyde oxime

Purity: 98% CAS No.: 1129-37-9 Molecular Formula: C7H6N2O3 Formula Weight: 166.14 Melting Point: 126-130° Beilstein Reference: 973581 UN No.: 2811 MDL No.: MFCD00007377

Formula: C₇H₆N₂O₃
MW: 166,14 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007377
CAS Number: 1129-37-9
EINECS: 214-445-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR