2-(Methylamino)benzotrifluoride 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-Methyl-2-(trifluoromethyl)aniline, 2-(Trifluoromethyl)-N-methylaniline
L09801.03
L09801.06
ALFAL09801.03EA
0
SGD
ALFAL09801.03
ALFAL09801.06
2-(Methylamino)benzotrifluoride 96%
2-(Methylamino)benzotrifluoride
Purity: 96%
CAS No.: 14925-10-1
Molecular Formula: C8H8F3N
Formula Weight: 175.16
Flash Point: 64°(147°F)
Refractive Index: 1.4860
Boiling Point: 177°
Beilstein Reference: 2641847
MDL No.: MFCD00052196
Formula:
C₈H₈F₃N MW: 175,15 g/mol Boiling Pt: 177 °C Density: 1,23 Flash Pt: 64 °C (147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052196 CAS Number: 14925-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...