4'-Bromobiphenyl-3-carboxylic acid 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4'-Bromo-[1,1'-biphenyl]-3-carboxylic acid
H51762.03
H51762.06
ALFAH51762.03EA
121
SGD
ALFAH51762.03
ALFAH51762.06
4'-Bromobiphenyl-3-carboxylic acid 95%
4'-Bromobiphenyl-3-carboxylic acid
Purity: 95%
CAS No.: 885951-66-6
Molecular Formula: C13H9BrO2
Formula Weight: 277.12
MDL No.: MFCD01318398
Formula:
C₁₃H₉BrO₂ MW: 277,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01318398 CAS Number: 885951-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...