4'-Methoxybiphenyl-3-carboxylic acid 95%

Supplier: Thermo Scientific
Warning

H51764.03 H51764.06
ALFAH51764.03EA 138 SGD
ALFAH51764.03 ALFAH51764.06
4'-Methoxybiphenyl-3-carboxylic acid 95%
4'-Methoxybiphenyl-3-carboxylic acid

Purity: 95% CAS No.: 725-05-3 Molecular Formula: C14H12O3 Formula Weight: 228.25 MDL No.: MFCD03424599

Formula: C₁₄H₁₂O₃
MW: 228,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424599
CAS Number: 725-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR