3-Fluoro-4-(trifluoromethoxy)benzaldehyde ≥97%
Supplier: Thermo Scientific
|
Warning
|
H26847.03
H26847.06
ALFAH26847.03EA
0
SGD
ALFAH26847.03
ALFAH26847.06
3-Fluoro-4-(trifluoromethoxy)benzaldehyde ≥97%
3-Fluoro-4-(trifluoromethoxy)benzaldehyde
Purity: 97%
CAS No.: 473917-15-6
Molecular Formula: C8H4F4O2
Formula Weight: 208.11
Refractive Index: 1.4455
Formula:
C₈H₄F₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00236275 CAS Number: 473917-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...