4,5-Difluoroanthranilic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-4,5-difluorobenzoic acid

L19950.03 L19950.06 L19950.14
ALFAL19950.03EA 0 SGD
ALFAL19950.03 ALFAL19950.06 ALFAL19950.14
4,5-Difluoroanthranilic acid 97%
4,5-Difluoroanthranilic acid

Purity: 97% CAS No.: 83506-93-8 Molecular Formula: C7H5F2NO2 Formula Weight: 173.12 Melting Point: 181-183° Beilstein Reference: 2834162

Formula: C₇H₅F₂NO₂
MW: 173,12 g/mol
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00077509
CAS Number: 83506-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR