1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Cyclen tetrahydrochloride

B22268.03 B22268.06
ALFAB22268.03EA 0 SGD
ALFAB22268.03 ALFAB22268.06
1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98%
1,4,7,10-Tetraazacyclododecane tetrahydrochloride

Purity: 98% CAS No.: 10045-25-7 Molecular Formula: C8H20N4.4HCl Formula Weight: 318.12 Odour: Odourless Melting Point: 290° UN No.: 3077 MDL No.: MFCD00068484

Formula: C₈H₂₄Cl₄N₄
Melting Pt: >290 °C
Storage Temperature: Ambient
MDL Number: MFCD00068484
CAS Number: 10045-25-7
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR