1,4,7,10-Tetraazacyclotridecane

Supplier: Thermo Scientific
Warning

H26671.MD H26671.03
ALFAH26671.MDEA 170 SGD
ALFAH26671.MD ALFAH26671.03
1,4,7,10-Tetraazacyclotridecane
1,4,7,10-Tetraazacyclotridecane

CAS No.: 295-14-7 Molecular Formula: C9H22N4 Formula Weight: 186.30 MDL No.: MFCD06410986

Formula: C₉H₂₂N₄
MW: 186,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410986
CAS Number: 295-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR