2-Benzyloxy-3-pyridineboronic acid pinacol ester 95%

Supplier: Thermo Scientific
Warning

H26928.MD H26928.03
ALFAH26928.MDEA 0 SGD
ALFAH26928.MD ALFAH26928.03
2-Benzyloxy-3-pyridineboronic acid pinacol ester 95%
2-Benzyloxy-3-pyridineboronic acid pinacol ester

Purity: 95% CAS No.: 1073371-81-9 Molecular Formula: C18H22BNO3 Formula Weight: 311.19 MDL No.: MFCD06798265

Formula: C₁₈H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD06798265
CAS Number: 1073371-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR