2-Methoxy-3-pyridineboronic acid pinacol ester ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Methoxypyridine-3-boronic acid pinacol ester, 2-Methoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H50060.03 H50060.MD
ALFAH50060.03EA 667 SGD
ALFAH50060.03 ALFAH50060.MD
2-Methoxy-3-pyridineboronic acid pinacol ester ≥97%
2-Methoxy-3-pyridineboronic acid pinacol ester

Purity: 97% CAS No.: 532391-31-4 Molecular Formula: C12H18BNO3 Formula Weight: 235.09 MDL No.: MFCD05663507

Formula: C₁₂H₁₈BNO₃
MW: 235,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663507
CAS Number: 532391-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR