2-Fluoro-3-methoxyphenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid

L19655.03 L19655.06
ALFAL19655.03EA 0 SGD
ALFAL19655.03 ALFAL19655.06
2-Fluoro-3-methoxyphenylboronic acid ≥97%
2-Fluoro-3-methoxyphenylboronic acid

Purity: 97% CAS No.: 352303-67-4 Molecular Formula: C7H8BFO3 Formula Weight: 169.95 Beilstein Reference: 9191510 MDL No.: MFCD02094728

Formula: C₇H₈BFO₃
MW: 169,95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094728
CAS Number: 352303-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR