α-Bromo-2,3,5,6-tetrafluorotoluene ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2,3,5,6-Tetrafluorobenzyl bromide, 2,3,5,6-Tetrafluorobenzylbromide

L01916.03 L01916.06
ALFAL01916.03EA 93 SGD
ALFAL01916.03 ALFAL01916.06
α-Bromo-2,3,5,6-tetrafluorotoluene ≥98%
α-Bromo-2,3,5,6-tetrafluorotoluene

Purity: 98+% CAS No.: 53001-73-3 Molecular Formula: C7H3BrF4 Formula Weight: 243.00 Refractive Index: 1.4885 Boiling Point: 79-80°/80mm UN No.: 3265 MDL No.: MFCD00013549

Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 79…80 °C (80 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00013549
CAS Number: 53001-73-3
EINECS: 258-289-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR