2-Bromo-4-(trifluoromethyl)aniline ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Amino-3-bromobenzotrifluoride, 4-Amino-3-bromo-α,α,α-trifluorotoluene, 2-Bromo-α,α,α-trifluoro-p-toluidine

A18528.03 A18528.06
ALFAA18528.03EA 0 SGD
ALFAA18528.03 ALFAA18528.06
2-Bromo-4-(trifluoromethyl)aniline ≥97%
2-Bromo-4-(trifluoromethyl)aniline

Purity: 97% CAS No.: 57946-63-1 Molecular Formula: C7H5BrF3N Formula Weight: 240.03 Flash Point: 110°(230°F) Refractive Index: 1.5240 Melting Point: 26-33° Boiling Point: 109-110°/10mm Beilstein Reference: 8139978 UN No.: 2811 MDL No.: MFCD00042150

Formula: C₇H₅BrF₃N
MW: 240,02 g/mol
Boiling Pt: 109…110 °C (10 mmHg)
Melting Pt: 26…33 °C
Density: 1,700
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042150
CAS Number: 57946-63-1
EINECS: 261-035-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR