2-Bromo-5-(trifluoromethyl)aniline ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Amino-4-bromobenzotrifluoride, 6-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-4-bromo-α,α,α-trifluorotoluene, 3-Amino-4-brom-α,α,α-trifluortoluol

A17810.03 A17810.06 A17810.14
ALFAA17810.03EA 66.6 SGD
ALFAA17810.03 ALFAA17810.06 ALFAA17810.14
2-Bromo-5-(trifluoromethyl)aniline ≥97%
2-Bromo-5-(trifluoromethyl)aniline

Purity: 97% CAS No.: 454-79-5 Molecular Formula: C7H5BrF3N Formula Weight: 240.03 Flash Point: 110°(230°F) Refractive Index: 1.5220 Boiling Point: 81-82°/5mm Beilstein Reference: 2094535 UN No.: 2810 MDL No.: MFCD00007637

Formula: C₇H₅BrF₃N
MW: 240,02 g/mol
Boiling Pt: 81…82 °C (5 mmHg)
Melting Pt: 47…49 °C
Density: 1,675
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007637
CAS Number: 454-79-5
EINECS: 207-227-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR