4-Bromo-2-(trifluoromethyl)aniline ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene
A12146.06
A12146.14
ALFAA12146.06EA
0
SGD
ALFAA12146.06
ALFAA12146.14
4-Bromo-2-(trifluoromethyl)aniline ≥98%
4-Bromo-2-(trifluoromethyl)aniline
Purity: 98%
CAS No.: 445-02-3
Molecular Formula: C7H5BrF3N
Formula Weight: 240.03
Flash Point: 107°(224°F)
Refractive Index: 1.5320
Boiling Point: 84-85°/5mm
Beilstein Reference: 2211504
UN No.: 2810
MDL No.: MFCD00064393
Formula:
C₇H₅BrF₃N MW: 240,02 g/mol Boiling Pt: 84…85 °C (5 mmHg) Density: 1,710 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064393 CAS Number: 445-02-3 EINECS: 207-150-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...