1-Phenyl-2-hexanone 95%
Supplier: Thermo Scientific
A10944.03
A10944.06
ALFAA10944.03EA
0
SGD
ALFAA10944.03
ALFAA10944.06
1-Phenyl-2-hexanone 95%
1-Phenyl-2-hexanone
Purity: 95%
CAS No.: 25870-62-6
Molecular Formula: C12H16O
Formula Weight: 176.26
Flash Point:
Formula:
C₆H₅CH₂C(O)(CH₂)₃CH₃ MW: 176,26 g/mol Boiling Pt: 153 °C (20 mmHg) Density: 0,940 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027240 CAS Number: 25870-62-6 EINECS: 247-306-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...