3-Amino-p-toluic acid 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Nilotinib Related Compound C, 3-Amino-4-methylbenzoic acid
A12674.06
A12674.14
A12674.22
ALFAA12674.06EA
0
SGD
ALFAA12674.06
ALFAA12674.14
ALFAA12674.22
3-Amino-p-toluic acid 99%
3-Amino-p-toluic acid
Purity: 99%
CAS No.: 2458-12-0
Molecular Formula: C8H9NO2
Formula Weight: 151.16
Melting Point: 164-168°
Beilstein Reference: 2082449
Formula:
C₈H₉NO₂ MW: 151,16 g/mol Boiling Pt: 363 °C (1013 hPa) Melting Pt: 164…168 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007742 CAS Number: 2458-12-0 EINECS: 219-543-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...